Learn More
Cayman Chemical Diquat (bromide), Purity: ≥95%, CAS: 85-00-7, 10 mg
SDP
Catalog No. 502932792
Encompass_Preferred
A bipyridylium herbicide; acts as an electron acceptor in the electron transport chain and is reoxidized by molecular oxygen, producing ROS; has been used to induce oxidative stress in vitro and in animal models; increases the production of ROS and decreases the mitochondrial membrane potential in mitochondria isolated from porcine intestine leading to mitophagy at 100 mg/kg
Catalog No. 50-293-2792 Supplier Cayman Chemical Supplier No. 2625110 MG
A bipyridylium herbicide; acts as an electron acceptor in the electron transport chain and is reoxidized by molecular oxygen, producing ROS; has been used to induce oxidative stress in vitro and in animal models; increases the production of ROS and decreases the mitochondrial membrane potential in mitochondria isolated from porcine intestine leading to mitophagy at 100 mg/kg