Learn More
Viagen Biotech Inc DIR PCR LYSIS RGNT(MOUSE TAIL)
SDP
Supplier:  Viagen Biotech Inc 102T
Encompass
DirectPCR Lysis Reagent (Mouse Tail) 100ml (Up to 500 Tails). The system is a single-tube system for rapid preparation of DNA from mouse tails. The patent-pending components allow the resulting DNA extracts to be compatible with genomic PCR for genotyping. Crude extracts of biological samples are not compatible with many molecular biology-grade reactions such as polymerase chain reaction (PCR), in part due to inhibitors contained in crude extracts. The DirectPCR reagents not only mediate the rapid lysis of biological samples but also contain inhibitors that effectively suppress the inhibitory activities of crude lysates for PCR amplification, while maximally maintaining the integrity of released genomic DNA. The patent-pending simple procedure completely eliminates any solution transfer or tube-opening steps, providing you with substantial extra time. Stable for 12 months at 4Deg C / Ships at room temperature.
Catalog No. NC9724951