Learn More
Enzo Life Sciences Direct 8-iso-PGF2alpha ELISA kit (5x96wells)
SDP
Catalog No. 502006892
Encompass_Preferred
Highly sensititive ELISA, measuring 8-iso-Prostaglandin F2α in plasma, serum and tissue. The measurement of F2-isoprostanes has emerged as one of the most reliable approaches to assess oxidative stress status in vivo, providing an important tool to explore the role of oxidative stress in the pathogenesis of human disease. The is a colorimetric competitive immunoassay kit for the quantitative determination of 8-iso-Prostaglandin F2α in biological fluids such as plasma, serum and tissue. To measure isoprostane in urine or culture supernatants, use the 8-iso-PGF2α EIA Kit. Alternative name: 8-iso-Prostaglandin F2alpha. Sensitivity: 40.0 pg/ml . Assay Time: <3 hours or Overnight + 45 min. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent . Content: GxR Microtiter plate, Conjugate, Antibody, Sample diluent, Neutralizing reagent, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.
Catalog No. 50-200-6892 Supplier Enzo Life Sciences Supplier No. ADI901091
Highly sensititive ELISA, measuring 8-iso-Prostaglandin F2α in plasma, serum and tissue. The measurement of F2-isoprostanes has emerged as one of the most reliable approaches to assess oxidative stress status in vivo, providing an important tool to explore the role of oxidative stress in the pathogenesis of human disease. The is a colorimetric competitive immunoassay kit for the quantitative determination of 8-iso-Prostaglandin F2α in biological fluids such as plasma, serum and tissue. To measure isoprostane in urine or culture supernatants, use the 8-iso-PGF2α EIA Kit. Alternative name: 8-iso-Prostaglandin F2alpha. Sensitivity: 40.0 pg/ml . Assay Time: <3 hours or Overnight + 45 min. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent . Content: GxR Microtiter plate, Conjugate, Antibody, Sample diluent, Neutralizing reagent, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.