Learn More
Cayman Chemical Direct Bilirubin Buffer, 8 mL
SDP
Catalog No. 502942512
Encompass_Preferred
Direct bilirubin buffer has been tested and formulated to work exclusively with Cayman’s Total and Direct Bilirubin Colorimetric Assay Kit (Item No. 701720).
Catalog No. 50-294-2512 Supplier Cayman Chemical Supplier No. 4004988 ML
Direct bilirubin buffer has been tested and formulated to work exclusively with Cayman’s Total and Direct Bilirubin Colorimetric Assay Kit (Item No. 701720).