Learn More
Biolegend Direct-Blot™ HRP anti-c-Myc
Supplier:  Biolegend 626803
Encompass_Preferred
Direct-Blot™ HRP anti-c-Myc [9E10]; Isotype: Mouse IgG1; Reactivity: Endogenous Human c-Myc; fusion protein with c-Myc tag (EQKLISEEDL, around 1.2kD) in all species; Apps: WB; Size: 100 uL
Catalog No. 50-208-6832