Learn More
Biolegend Direct-Blot™ HRP anti-ERK1/2 Phospho (Thr202/Tyr204)
Supplier:  Biolegend 675506
Encompass_Preferred
Direct-Blot™ HRP anti-ERK1/2 Phospho (Thr202/Tyr204) [4B11B69]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: WB; Size: 25 uL
Catalog No. 50-208-7345