Learn More
Biolegend Direct-Blot™ HRP anti-GST
Supplier:  Biolegend 640804
Encompass_Preferred
Direct-Blot™ HRP anti-GST [P1A12]; Isotype: Mouse IgG2b, κ; Reactivity: GST tagged protein; Apps: WB; Size: 25 uL
Catalog No. 50-208-6876