Learn More
Biolegend Direct-Blot™ HRP anti-HSP60
Supplier:  Biolegend 681503
Encompass_Preferred
Direct-Blot™ HRP anti-HSP60 [P83G8]; Isotype: Mouse IgG1; Reactivity: Human, Mouse, Rat; Apps: WB; Size: 100 uL
Catalog No. 50-208-7433