Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Direct blue 14; Trypan blue | 72-57-1 | 39.0% | 10 G
SDP

Catalog No. 5000409748
Encompass_Preferred

Diphenyl Blue (Trypan Blue) is a cell-active dye, most commonly used for identifying dead cells. It is frequently employed to test cell membrane integrity and cell viability. Diphenyl Blue staining is a method used in tissue and cell culture. Deactivated cells or those with incomplete cell membranes are stained blue by Diphenyl Blue. Normal living cells with intact cell membranes, however, reject Diphenyl Blue and do not stain blue. Macrophages can phagocytose Diphenyl Blue, allowing it to be used as a living stain for these cells.

  • Cell active dye
  • Identifies dead cells
  • Tests cell membrane integrity and cell viability
  • Can be used as a living stain for macrophages
  • Molecular Weight: 960.81
  • Formula: C34H24N6Na4O14S4
  • Appearance: Solid
  • Color: Light brown to black

Catalog No. 50-004-09748 Supplier Medchemexpress LLC Supplier No. HYD097010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diphenyl Blue (Trypan Blue) is a cell-active dye, most commonly used for identifying dead cells. It is frequently employed to test cell membrane integrity and cell viability. Diphenyl Blue staining is a method used in tissue and cell culture. Deactivated cells or those with incomplete cell membranes are stained blue by Diphenyl Blue. Normal living cells with intact cell membranes, however, reject Diphenyl Blue and do not stain blue. Macrophages can phagocytose Diphenyl Blue, allowing it to be used as a living stain for these cells.

  • Cell active dye
  • Identifies dead cells
  • Tests cell membrane integrity and cell viability
  • Can be used as a living stain for macrophages
  • Molecular Weight: 960.81
  • Formula: C34H24N6Na4O14S4
  • Appearance: Solid
  • Color: Light brown to black

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.