Learn More
Phenomenex Inc for valves w/ port size of <150 microns, Hand tighten (MAX 25,000psi)
Catalog No. 502400932 Shop All Phenomenex Products
Encompass_Preferred
Direct Connect Fittings, Knurled main body, Connects 360 m AN0-1012
Catalog No. 50-240-0932 Supplier Phenomenex Inc Supplier No. AN01012
Direct Connect Fittings, Knurled main body, Connects 360 m AN0-1012