Learn More
Research Products International Corp Direct Thermal Cryo-Tags, 1.05 x 0.50, White, 1000/rl
SDP
Supplier:  Research Products International Corp 249400
Encompass
Cryogenic labels for direct thermal printers. Direct Thermal Cryo-Tags are formatted to work quickly and easily with the Dymo Labelwriter and most direct thermal printers. Quickly label your samples with sharp text, clear barcodes or a combination of both! Cryogenic labels are suitable for a variety of applications. Recommended Temperature: -196°C to +70°C
Catalog No. 50-197-8221