Learn More
Arbor Assays Direct Cyclic GMP (cGMP) ELISA Kit (1 Plate)
SDP
Supplier:  Arbor Assays K065H1
Encompass_Preferred
The DetectX cGMP Immunoassay Kits - Improved Sensitivity is designed to measure cGMP directly and conveniently lyse, stabilize and measure in one step.
Catalog No. 50-227-8497