Learn More
MSC TPI Exhaust Fan, 10 in. Blade, 3-Speed, 1/12 HP, 1 ea.
Catalog No. 5000507155
Encompass
This 10 in. direct drive exhaust fan is engineered for industrial and commercial ventilation requirements. Featuring a robust 1/12 HP motor, this unit provides reliable air circulation with a maximum flow rate of 680 CFM. Designed for direct wall mounting, it includes a wire guard for safety and is built with durable materials, including an aluminum blade for long-term performance.
  • Three-speed motor settings for adjustable airflow
  • Direct drive operation with a maximum speed of 1,560 RPM
  • Compatible with optional external shutters
  • Includes factory-lubricated motor bearings
  • Equipped with a convenient pull chain switch
  • Junction box included for direct wiring applications
  • Designed for 120 V, 1-phase power supply
Catalog No. 50-005-07155 Supplier MSC Supplier No. 77994739
Add to Cart
Add to Cart
This 10 in. direct drive exhaust fan is engineered for industrial and commercial ventilation requirements. Featuring a robust 1/12 HP motor, this unit provides reliable air circulation with a maximum flow rate of 680 CFM. Designed for direct wall mounting, it includes a wire guard for safety and is built with durable materials, including an aluminum blade for long-term performance.
  • Three-speed motor settings for adjustable airflow
  • Direct drive operation with a maximum speed of 1,560 RPM
  • Compatible with optional external shutters
  • Includes factory-lubricated motor bearings
  • Equipped with a convenient pull chain switch
  • Junction box included for direct wiring applications
  • Designed for 120 V, 1-phase power supply