Learn More
WELCH MATERIALS INC Direct Connect Analytical Guard Cartridge Holder(φ: 4.6mm, 7000psi)
Catalog No. NC2052541
Encompass_Preferred
Direct Connect Analytical Guard Cartridge Holder(φ: 4.6mm, 7000psi), compatible with Parker, Valco, Waters columns
Catalog No. NC2052541 Supplier WELCH MATERIALS INC Supplier No. 0080801108
Direct Connect Analytical Guard Cartridge Holder(φ: 4.6mm, 7000psi), compatible with Parker, Valco, Waters columns