Learn More
Pointe Scientific Direct Hemoglobin A1c (HbA1c)
Catalog No. 22666104 Shop All Pointe Scientific Products
Change view
Click to view available options
Type:
HbA1c Control
HbA1c Lyse
Hemoglobin A1c Calibrator Set
Quantity:
4 x 0.5 mL
1 x 500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
22-666-104 HbA1c Lyse 1 x 500 mL
22-666-102 Hemoglobin A1c Calibrator Set 4 x 0.5 mL
22-666-103 HbA1c Control 4 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-666-104 Supplier Pointe Scientific Supplier No. H7541LYS
Add to Cart
Add to Cart
Single Channel Method
•    A latex-enhanced immunoturbidimetric assay for the automated determination of HbA1c
•    For use with routine chemistry analyzers
•    Calibrators referenced to NGSP value (assigned HbA1c frozen whole blood)
•    Normal and elevated controls are available
•    Reagents are liquid stable for cost-effective use of the entire kit
Specifications
Certifications/Compliance NGSP
Type HbA1c Lyse
For Use With (Application) Automated HbA1c Determination
Quantity 1 x 500 mL