Learn More
Creative Biomart Desirudin
Catalog No. 502084065
Encompass_Preferred
The product is a direct inhibitor of human thrombin. It has a protein structure that is similar to that of hirudin, the naturally occurring anticoagulant present in the peripharyngeal glands in the medicinal leech, Hirudo medicinalis. Hirudin is a single polypeptide chain of 65 amino acids residues and contains three disulfide bridges. The product has a chemical formula of C287H440N80O110S6 with a molecular weight of 6963.52.
Catalog No. 50-208-4065 Supplier Creative Biomart Supplier No. THP0014
The product is a direct inhibitor of human thrombin. It has a protein structure that is similar to that of hirudin, the naturally occurring anticoagulant present in the peripharyngeal glands in the medicinal leech, Hirudo medicinalis. Hirudin is a single polypeptide chain of 65 amino acids residues and contains three disulfide bridges. The product has a chemical formula of C287H440N80O110S6 with a molecular weight of 6963.52.