Learn More
LAMDA BIOTECH INC DIRECT-2X PCR MIX 1ml
Catalog No. NC3726387
Encompass_Preferred
Direct-PCR™ 2X MasterMix is a specialized PCR formulation engineered for direct amplification from unpurified DNA samples. This ready-to-use master mix combines Taq Plus™ DNA polymerase with optimized buffer components for efficient genotyping applications.
Catalog No. NC3726387 Supplier LAMDA BIOTECH INC Supplier No. D300100MIX
Direct-PCR™ 2X MasterMix is a specialized PCR formulation engineered for direct amplification from unpurified DNA samples. This ready-to-use master mix combines Taq Plus™ DNA polymerase with optimized buffer components for efficient genotyping applications.