Learn More
LAMDA BIOTECH INC DIRECT-PCR Geno DNA Prep
Catalog No. NC3724381
Encompass_Preferred
The Direct™ PCR Genotyping DNA Prep Solution is a specialized reagent designed for rapid genomic DNA preparation for direct PCR applications. This solution streamlines genotyping workflows by enabling DNA extraction without traditional purification steps.
Catalog No. NC3724381 Supplier LAMDA BIOTECH INC Supplier No. D3001000SOLN
The Direct™ PCR Genotyping DNA Prep Solution is a specialized reagent designed for rapid genomic DNA preparation for direct PCR applications. This solution streamlines genotyping workflows by enabling DNA extraction without traditional purification steps.