Learn More
LAMDA BIOTECH INC DIRECT-PCR KIT: Seeds 1000rxn
Catalog No. NC3724384
Encompass_Preferred
A direct PCR system designed for genomic DNA extraction and amplification from seed samples. Contains optimized reagents for processing multiple seed types without separate DNA purification steps.
Catalog No. NC3724384 Supplier LAMDA BIOTECH INC Supplier No. D3031000
A direct PCR system designed for genomic DNA extraction and amplification from seed samples. Contains optimized reagents for processing multiple seed types without separate DNA purification steps.