Learn More
LaMotte™ Direct Reading Titrator
SDPSCI_ED
Catalog No. S72342 Shop All LaMotte Company Products
Change view
Click to view available options
Volume (Metric) Sample:
1 mL
Catalog No. Volume (Metric) Sample
S72342 1 mL
1 options
Catalog No. S72342 Supplier LaMotte™ Supplier No. 0382
Add to Cart
Add to Cart
Replacement for glass direct reading titrators
  • Chemically resistant plastic
  • One-piece plunger
  • Special precision tip
  • Range: 0 to 200
Specifications
Type Direct Reading Titrator
For Use With (Application) Determine the concentration of a substance in a sample solution
For Use With (Equipment) LaMotte water quality monitoring kits, including LaMotte DO kits- LaMotte DO kit, LaMotte deluxe water quality education outfit, LaMotte saltwater aquaculture test kit and LaMotte freshwater aquaculture test kit
Includes One-piece plunger and special precision tip
Material Plastic
Volume (Metric) Sample 1 mL
WARNING: Cancer - www.P65Warnings.ca.gov