Learn More
Cayman Chemical Direct Red 80, CAS: 2610-10-8, 10 g
SDP
Catalog No. 502916214
Encompass_Preferred
An anionic hexasulfonic acid dye; abs = 540 nm; has been used as a crosslinking dye for collagen or amyloids, as well as a dye for fabrics
Catalog No. 50-291-6214 Supplier Cayman Chemical Supplier No. 3953510 G
An anionic hexasulfonic acid dye; abs = 540 nm; has been used as a crosslinking dye for collagen or amyloids, as well as a dye for fabrics