Learn More
Selleck Chemical LLC DIRECT RED 80 E2905-25MG
SDP
Catalog No. 5000040374
Encompass_Preferred
Direct Red 80 (Sirius Red) is a polyazo dye used principally in staining methods for collagen and amyloid Direct Red 80 does not release benzidine upon degradation and is safer than many traditional direct dyes
Catalog No. 50-000-40374 Supplier Selleck Chemical LLC Supplier No. E290525MG
Direct Red 80 (Sirius Red) is a polyazo dye used principally in staining methods for collagen and amyloid Direct Red 80 does not release benzidine upon degradation and is safer than many traditional direct dyes