Learn More
Description
Direct Red 80 (C. I. 35780) is an extremely hydrophilic, very large tetrakisazo hexasulfonated anionic dye that has a large, planar aromatic system. It is also known as Sirius red F3B, chlorantine fast red 7BLN or 5BRL, durazol brilliant red B, pontamine fast red 7BNL, and sirius fast red.
Specifications
Specifications
| CAS | 2610-10-8 |
| Molecular Weight (g/mol) | 1373.07 g/mol |
| Color | Dark Brown |
| Quantity | 25 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.