Learn More
Alpco Ltd Direct Saliva Melatonin ELISA
SDP
Supplier:  Alpco Ltd 12EKDSMU
Encompass
The Direct Saliva Melatonin ELISA kit is intended for the quantitative determination of melatonin in human saliva samples. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2284499