Learn More
Andwin Scientific DIRECT THERMAL CRYO Tage 27 x 13 1,000 pack
SDP
Catalog No. 50727690
Encompass_Preferred
Compatible with direct thermal printers, the 27X13mm Cryo-Tags are used on 0.5ml tubes. They are temperature resistant from -196-70 deg.C and freezable in liquid nitrogen. It comes in 1000 a pack.
Catalog No. 50-727-690 Supplier Andwin Scientific Supplier No. 775771
Compatible with direct thermal printers, the 27X13mm Cryo-Tags are used on 0.5ml tubes. They are temperature resistant from -196-70 deg.C and freezable in liquid nitrogen. It comes in 1000 a pack.