Learn More
Alkali Scientific Direct Thermal Cryo Label, 22x22mm for Microscope Slides, 1000 per Roll, 1/Roll
SDP
Catalog No. 5000661980
Encompass_Preferred
Cryogenic, waterproof labels suitable for cryovials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other types of containers for storage in liquid and vapor phase nitrogen ( 196°C), laboratory freezers ( 120ºC, 80ºC, 20ºC), and transportation on dry ice. Comprised of a thin thermoplastic film and a special cryogenic adhesive, these durable labels do not become brittle or crack, and have a strong affinity for polyethylene, polypropylene and other plastic and glass surfaces. They also resist smearing without additional lamination or wrap around. Available in white or a variety of colors upon request. Supplied in roll format and are compatible with all major brands of direct thermal barcode printers, such as Zebra, Intermec, Citizen, Cognitive, Printronix, Datamax O`Neil, SATO and others. They can also be written on with our cryo or permanent markers. The labels are resistant to standard ethylene oxide gas sterilization, consisting of a 60 minute exposure to 75
Catalog No. 50-006-61980 Supplier Alkali Scientific Supplier No. LDTC7878W
Cryogenic, waterproof labels suitable for cryovials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other types of containers for storage in liquid and vapor phase nitrogen ( 196°C), laboratory freezers ( 120ºC, 80ºC, 20ºC), and transportation on dry ice. Comprised of a thin thermoplastic film and a special cryogenic adhesive, these durable labels do not become brittle or crack, and have a strong affinity for polyethylene, polypropylene and other plastic and glass surfaces. They also resist smearing without additional lamination or wrap around. Available in white or a variety of colors upon request. Supplied in roll format and are compatible with all major brands of direct thermal barcode printers, such as Zebra, Intermec, Citizen, Cognitive, Printronix, Datamax O`Neil, SATO and others. They can also be written on with our cryo or permanent markers. The labels are resistant to standard ethylene oxide gas sterilization, consisting of a 60 minute exposure to 75