Learn More
Alkali Scientific ASI™ Direct Thermal Cryo Label, 24x13mm & 9.5mm Dot for 1.5mL Tubes/Vials, 500 Sets, 1/Roll, Orange
SDP
Catalog No. 503028841
Encompass_Preferred
DIRECT THERMAL CRYO LABEL 24x13mm & 9.5mm Dot for 1.5ml Tubes/Vials We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Labels Per case: 500 sets/1 Roll/Cs
Catalog No. 50-302-8841 Supplier Alkali Scientific Supplier No. LDTC11238O
DIRECT THERMAL CRYO LABEL 24x13mm & 9.5mm Dot for 1.5ml Tubes/Vials We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Labels Per case: 500 sets/1 Roll/Cs