Learn More
Alkali Scientific ASI™ Direct Thermal Cryo Label, 27x13mm for Cryogenic Vials, 1000 per Roll, 1/Roll, Yellow
SDP
Catalog No. 503027062
Encompass_Preferred
DIRECT THERMAL CRYO LABEL 27x13mm for Cryogenic Vials We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Labels Per case: 1000, 1 roll/cs
Catalog No. 50-302-7062 Supplier Alkali Scientific Supplier No. LDTC112Y
DIRECT THERMAL CRYO LABEL 27x13mm for Cryogenic Vials We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Labels Per case: 1000, 1 roll/cs