Learn More
Alkali Scientific ASI™ Direct Thermal Cryo Label, 33x13mm & 9.5mm Dot for 2.0mL Tubes/Vials, 500 Sets, 1/Roll, White
SDP
Catalog No. 503027193
Encompass_Preferred
DIRECT THERMAL CRYO LABEL 33x13mm & 9.5mm Dot for 2.0ml Tubes/Vials We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Labels Per roll: 500, 1 Roll/cs
Catalog No. 50-302-7193 Supplier Alkali Scientific Supplier No. LDTC1141238W
DIRECT THERMAL CRYO LABEL 33x13mm & 9.5mm Dot for 2.0ml Tubes/Vials We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Labels Per roll: 500, 1 Roll/cs