Learn More
Alkali Scientific ASI™ Direct Thermal Cryo Label, 38x13mm & 9.5mm Dot for 2.0mL Tubes/Vials, 500 Sets, 1/Roll, Yellow
SDP
Catalog No. 503028850
Encompass_Preferred
DIRECT THERMAL CRYO LABEL 38x13mm & 9.5mm Dot for 2.0ml Tubes/Vials We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Packing: 500 sets/ 1 Roll/Cs
Catalog No. 50-302-8850 Supplier Alkali Scientific Supplier No. LDTC1121238Y
DIRECT THERMAL CRYO LABEL 38x13mm & 9.5mm Dot for 2.0ml Tubes/Vials We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Packing: 500 sets/ 1 Roll/Cs