Learn More
Alkali Scientific Direct Thermal Cryo Label, 43x19mm for Lg Vials/Tubes, 500 per Roll, 1/Roll
SDP
Catalog No. 5000662069
Encompass_Preferred
Cryogenic, waterproof labels suitable for cryovials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other types of containers for storage in liquid and vapor phase nitrogen ( 196°C), laboratory freezers ( 120ºC, 80ºC, 20ºC), and transportation on dry ice. Comprised of a thin thermoplastic film and a special cryogenic adhesive, these durable labels do not become brittle or crack, and have a strong affinity for polyethylene, polypropylene and other plastic and glass surfaces. They also resist smearing without additional lamination or wrap around. Available in white or a variety of colors upon request. Supplied in roll format and are compatible with all major brands of direct thermal barcode printers, such as Zebra, Intermec, Citizen, Cognitive, Printronix, Datamax O`Neil, SATO and others. They can also be written on with our cryo or permanent markers. The labels are resistant to standard ethylene oxide gas sterilization, consisting of a 60 minute exposure to 75
Catalog No. 50-006-62069 Supplier Alkali Scientific Supplier No. LDTC34169W
Cryogenic, waterproof labels suitable for cryovials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other types of containers for storage in liquid and vapor phase nitrogen ( 196°C), laboratory freezers ( 120ºC, 80ºC, 20ºC), and transportation on dry ice. Comprised of a thin thermoplastic film and a special cryogenic adhesive, these durable labels do not become brittle or crack, and have a strong affinity for polyethylene, polypropylene and other plastic and glass surfaces. They also resist smearing without additional lamination or wrap around. Available in white or a variety of colors upon request. Supplied in roll format and are compatible with all major brands of direct thermal barcode printers, such as Zebra, Intermec, Citizen, Cognitive, Printronix, Datamax O`Neil, SATO and others. They can also be written on with our cryo or permanent markers. The labels are resistant to standard ethylene oxide gas sterilization, consisting of a 60 minute exposure to 75