Learn More
Alkali Scientific ASI™ Direct Thermal Cryo Label, 51x19mm for Lg Tubes/Racks/Boxes, 500 per Roll, 1/Roll, Orange
SDP
Catalog No. 503029438
Encompass_Preferred
DIRECT THERMAL CRYO LABEL 51x19mm for Lg Tubes/Racks/Boxes We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Labels Per roll: 500, 1 Roll/cs
Catalog No. 50-302-9438 Supplier Alkali Scientific Supplier No. LDTC342O
DIRECT THERMAL CRYO LABEL 51x19mm for Lg Tubes/Racks/Boxes We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C). Labels Per roll: 500, 1 Roll/cs