Learn More
GA International Direct Thermal Heat-Resistant Labels (Patent Pending) 1" x 3" / 25.4mm x 76.2mm Rectangle White From -80°C to +95°C (-112°F to +203°F)
Catalog No. 5000570857
Encompass_Preferred
These heat-resistant labels are compatible with most common direct thermal printers. They can withstand elevated temperatures up to +95°C (+203°F) under both dry and wet conditions, which allows them to be used for different laboratory tasks that include heat inactivation and high heat treatments*. The primary quality of these high heat labels is their ability to remain readable after exposure to hot conditions, in contrast with standard direct thermal labels that turn entirely dark during the heating process. These permanent labels are suitable for identifying various laboratory labware and equipment composed of different materials, including metal, glass, and plastic. Given that these labels are also resistant to low temperatures (-80°C/ -112°F), they can be utilized for freezing applications and storage.*Please note that some minor label darkening/mottling may occur after standard autoclave conditions, boiling water, or temperature exposure exceeding the recommended limit. Prior t
Catalog No. 50-005-70857 Supplier GA International Supplier No. DH218C31WH
These heat-resistant labels are compatible with most common direct thermal printers. They can withstand elevated temperatures up to +95°C (+203°F) under both dry and wet conditions, which allows them to be used for different laboratory tasks that include heat inactivation and high heat treatments*. The primary quality of these high heat labels is their ability to remain readable after exposure to hot conditions, in contrast with standard direct thermal labels that turn entirely dark during the heating process. These permanent labels are suitable for identifying various laboratory labware and equipment composed of different materials, including metal, glass, and plastic. Given that these labels are also resistant to low temperatures (-80°C/ -112°F), they can be utilized for freezing applications and storage.*Please note that some minor label darkening/mottling may occur after standard autoclave conditions, boiling water, or temperature exposure exceeding the recommended limit. Prior t