Learn More
PDC Direct Thermal IR Paper Permanent, 1300/Roll, 8 Rolls/Case
Catalog No. 21235876
Change view
Click to view available options
Quantity:
1300/Roll, 8 Rolls/Cs.
Catalog No. Quantity
21-235-876 1300/Roll, 8 Rolls/Cs.
1 options
Catalog No. 21-235-876 Supplier PDC Supplier No. THERMIRD19
Add to Cart
Add to Cart
Ideal for healthcare and industrial applications requiring durable, reliable labeling.
  • Compatible with Eltron LP, TLP Series, and other thermal printers
  • White surface provides high contrast for crisp, legible printing
  • Strong adhesive keeps labels securely in place on various surfaces
  • Perforated design allows easy separation and handling
  • Suitable for clinical, healthcare, inventory, and asset tracking applications
  • Infrared (IR) paper technology improves print quality and durability
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1300/Roll, 8 Rolls/Cs.
Dimensions (L x W) 2 1/2 x 1 in.
Length (English) 1 in.
Width (English) 2.5 in.
Width (Metric) 63.5 mm