Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caresfield Direct Thermal Label, 1250/Roll, 8 Rolls/Box
SDP

Catalog No. 21222097
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-222-097 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-222-097 Supplier Caresfield Supplier No. 86716
Only null left
Add to Cart
Add to Cart

1.25 x 3 in. Direct Thermal Label

  • Ideal for quick labeling applications in healthcare, labs, logistics, inventory, and general purpose identification.

Specifications

Adhesive Type Acrylic
Coating None
Color White
Finish Matte White
Format Label
Material Plastic
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1250/Roll, 8 Rolls/Box
Dimensions (LxW) 1.25 x 4 in.
Length (English) 1.25 in.
Width (English) 3 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.