Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caresfield Direct Thermal Label, 2340/Roll, 8 Rolls/Cs.
SDP

Catalog No. 21222105
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-222-105 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-222-105 Supplier Caresfield Supplier No. 83712
Only null left
Add to Cart
Add to Cart

2 x 1 in. Direct Thermal Label 1 in. Core Wound Out

  • Ideal for shipping, barcode, inventory, shelf, and product identification labeling.

Specifications

Autoclavable Not Autoclavable
Color Capability Monochrome
Adhesive Type Acrylic, Pressure-sensitive, Permanent
Coating None
Color White
Finish Matte White
Format Label
Material Plastic
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Languages No Text
Latex Free Yes
Quantity 2340/Roll, 8 Rolls/Cs.
Resolution 203 dpi
Shape Rectangle with Rounded Corners
Dimensions (LxW) 2 x 1 in.
Length (English) 2 in.
Width (English) 1 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.