Learn More
PDC Direct Thermal Label, 250 Labels/Roll, 8 Rolls/Box
Catalog No. 21235832
Change view
Click to view available options
Quantity:
250 Labels/Roll, 8 Rolls/Box
Catalog No. Quantity
21-235-832 250 Labels/Roll, 8 Rolls/Box
1 options
Catalog No. 21-235-832 Supplier PDC Supplier No. TD751142
Add to Cart
Add to Cart
Permanent adhesive labels designed for Epic and Cerner compatibility, ensuring accurate blood tube identification with V-Notch alignment.
  • Engineered for seamless use with Epic and Cerner EHR systems to support efficient lab workflows
  • Permanent adhesive ensures labels stay securely on tubes and containers
  • V-Notch™ design allows precise alignment with color-coded notches for consistent placement
  • Open window feature enables quick verification of sample quality and fill levels
  • Improves barcode scanning accuracy and readability for faster processing
  • Helps reduce redraws, misidentification, and test delays in clinical environments
  • Supports operational efficiency and enhances clinician and patient satisfaction
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 250 Labels/Roll, 8 Rolls/Box
Dimensions (L x W) 2 x 1 1/4 in.
Length (English) 2 in.
Width (English) 1.25 in.
Width (Metric) 31.75 mm