Learn More
PDC Direct Thermal Label, Green Border, 350 Labels/Roll, 36 Rolls/Case
Catalog No. 21235786
Change view
Click to view available options
Quantity:
350 Labels/Roll, 36 Rolls/Cs.
Catalog No. Quantity
21-235-786 350 Labels/Roll, 36 Rolls/Cs.
1 options
Catalog No. 21-235-786 Supplier PDC Supplier No. LDQL220GB
Add to Cart
Add to Cart
Green-bordered direct thermal labels provide clear, color-coded identification for healthcare and laboratory use.
  • Designed for sharp, ink-free printing with consistent results
  • Green border enhances visibility and supports color-coded labeling workflows
  • Black sensor bar ensures reliable printer detection and alignment
  • Permanent adhesive secures labels firmly to vials, tubes, and containers
  • Compact 3/4 in. core works with small-format dispensers for easy application
  • Bulk supply with 350 labels per roll and 36 rolls per case supports high-volume needs
Specifications
Adhesive Type Permanent
Coating None
Color White, Green Border
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 350 Labels/Roll, 36 Rolls/Cs.
Dimensions (L x W) 2 x 1 in.
Length (English) 2 in.
Width (English) 1 in.
Width (Metric) 25.4 mm