Learn More
PDC Direct Thermal Label, Paper, 1200 Labels/Roll, 8 Rolls/Box
Catalog No. 21235843
Change view
Click to view available options
Quantity:
1200 Labels/Roll, 8 Rolls/Box
Catalog No. Quantity
21-235-843 1200 Labels/Roll, 8 Rolls/Box
1 options
Catalog No. 21-235-843 Supplier PDC Supplier No. TDSQBLSSL
  • $308.00
    $230.65 / Case of 8
    Save $77.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Medical-grade labels with permanent adhesive, compatible with Epic, Misys, and Sunquest systems, supporting accurate multi-specimen collection.
  • Designed for use with Epic Beaker/Lab Module, Misys, and Sunquest EHR systems to streamline lab workflows
  • Black sensor bar ensures reliable printer detection and accurate alignment
  • Multi-piece format supports multiple specimen collection for efficient processing
  • Permanent adhesive keeps labels securely attached to tubes, vials, and containers
  • Direct thermal technology allows easy printing without ribbons or ink
  • Enhances workflow accuracy, reduces labeling errors, and supports patient safety
  • Bulk packaging provides ample supply for frequent laboratory and clinical use
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1200 Labels/Roll, 8 Rolls/Box
Dimensions (L x W) 4 1/8x1 3/16 in.
Length (English) 4.125 in.
Width (English) 1.1875 in.
Width (Metric) 30.1625 mm