Learn More
PDC Direct Thermal Label, Paper, 1200 Labels/Roll, 8 Rolls/Box
Catalog No. 21235844
Change view
Click to view available options
Quantity:
1200 Labels/Roll, 8 Rolls/Box
Catalog No. Quantity
21-235-844 1200 Labels/Roll, 8 Rolls/Box
1 options
Catalog No. 21-235-844 Supplier PDC Supplier No. TDSQBLSTR
  • $377.00
    $281.65 / Case of 8
    Save $95.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Perforated lab labels with permanent adhesive, compatible with Epic, Misys, and Sunquest for accurate multi-specimen labeling.
  • Designed for Epic, Misys, and Sunquest EHR systems to streamline lab workflows
  • Black sensor bar ensures reliable printing and accurate alignment
  • Red border on multi-piece labels provides clear visual identification
  • Perforated design allows easy separation and handling for multiple specimens
  • Permanent adhesive keeps labels securely attached to vials, tubes, and containers
  • Supports patient safety, improves specimen accuracy, and reduces labeling errors
  • Direct thermal printing enables jam-free, efficient label production for high-volume labs
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1200 Labels/Roll, 8 Rolls/Box
Dimensions (L x W) 4 1/8 x 1 3/16 in.
Length (English) 4.125 in.
Width (English) 1.1875 in.
Width (Metric) 30.1625 mm