Learn More
PDC Direct Thermal Label, Paper, 4300 Labels/Roll, 2 Rolls/Box
Catalog No. 21235842
Change view
Click to view available options
Quantity:
4300 Labels/Roll, 2 Rolls/Box
Catalog No. Quantity
21-235-842 4300 Labels/Roll, 2 Rolls/Box
1 options
Catalog No. 21-235-842 Supplier PDC Supplier No. TDSQ4
  • $208.00
    $155.65 / Case of 2
    Save $52.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Reliable direct thermal labels with permanent adhesive and sensor bar, compatible with Epic, Misys, and Sunquest for secure specimen ID.
  • Compatible with Epic, Misys, and Sunquest EHR systems for seamless lab workflow integration
  • Permanent adhesive ensures labels remain securely attached to specimens and containers
  • Black sensor bar provides consistent printer detection and accurate alignment
  • Multi-piece label design supports multiple specimen collection efficiently
  • Medical-grade paper ensures reliable readability and barcode scanning in clinical labs
  • Enhances workflow accuracy, reduces errors, and improves patient safety
  • Quick, jam-free printing supports operational efficiency and satisfaction for lab staff
  • Bulk packaging provides sufficient supply for high-volume labeling needs
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 4300 Labels/Roll, 2 Rolls/Box
Dimensions (L x W) 4 1/8 x 1 3/16 in.
Length (English) 4.125 in.
Width (English) 1.1875 in.
Width (Metric) 30.1625 mm