Learn More
PDC Direct Thermal Paper Label, 500 Labels/Roll
Catalog No. 21235807
Change view
Click to view available options
Quantity:
500 Labels/Roll
Catalog No. Quantity
21-235-807 500 Labels/Roll
1 options
Catalog No. 21-235-807 Supplier PDC Supplier No. LDWHP764
Add to Cart
Add to Cart
White labels with strong permanent adhesion ensure reliable identification, supporting accuracy and safety in clinical and laboratory workflows.
  • Permanent adhesive keeps labels securely in place for critical healthcare and lab use
  • White face stock delivers crisp, clear printing for easy readability
  • Larger format supports detailed specimen or patient information labeling
  • Compatible with standard dispensers for smooth application
  • Durable design ensures long-lasting performance in demanding environments
  • Packaged for consistent supply in clinical and laboratory settings
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 500 Labels/Roll
Dimensions (L x W) 4 x 5 1/2 in.
Length (English) 4 in.
Width (English) 5.5 in.
Width (Metric) 139.7 mm