Learn More
PDC Direct Thermal Paper Label Permanent, 1300/Roll, 8 Rolls/Case
Catalog No. 21235852
Change view
Click to view available options
Quantity:
1300/Roll, 8 Rolls/Cs.
Catalog No. Quantity
21-235-852 1300/Roll, 8 Rolls/Cs.
1 options
Catalog No. 21-235-852 Supplier PDC Supplier No. THERMD19
Add to Cart
Add to Cart
Designed for Eltron LP and TLP printers, these labels provide clear printing, reliable adhesion, and consistent performance in healthcare use.
  • Compatible with Eltron LP, TLP series, and other printers
  • Permanent adhesive ensures reliable label adhesion
  • Direct thermal printing eliminates need for ink or ribbons
  • Suitable for diverse labeling needs
  • Corner radius prevents peeling and curling
  • Perforated design allows easy label separation
  • Fits standard printer requirements
  • Glove-friendly adhesive options for healthcare environments
  • Packed for long-lasting supply
  • Multi-purpose use supports specimen, inventory, and general labeling
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1300/Roll, 8 Rolls/Cs.
Dimensions (L x W) 2 1/2 x 1 in.
Length (English) 2.5 in.
Width (English) 1 in.
Width (Metric) 25.4 mm