Learn More
PDC Direct Thermal Paper Label Permanent, 1300/Roll, 8 Rolls/Case
Catalog No. 21235856
Change view
Click to view available options
Quantity:
1300/Roll, 8 Rolls/Cs.
Catalog No. Quantity
21-235-856 1300/Roll, 8 Rolls/Cs.
1 options
Catalog No. 21-235-856 Supplier PDC Supplier No. THERMD24
Add to Cart
Add to Cart
Compatible with Eltron LP and TLP printers, these labels provide strong adhesion, sharp printing, and versatile labeling performance.
  • Compatible with Eltron LP, TLP Series, and other widely used printers for seamless functionality
  • Permanent adhesive ensures reliable and long-lasting labels that stay firmly in place
  • Direct thermal printing requires no ink or ribbon, helping to simplify processes and reduce costs
  • Designed for a variety of identification and labeling needs across industries
  • Rounded corner design prevents peeling and curling, ensuring smooth application
  • Perforated layout allows easy label separation, saving time and improving workflow
  • Optimized roll format supports standard printer compatibility for consistent output
  • Glove-friendly adhesive ensures safe and easy handling in healthcare and laboratory environments
  • Bulk packaging provides continuous supply for high-volume labeling requirements
  • Multi-purpose solution suitable for asset tracking, specimen labeling, and general applications
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1300/Roll, 8 Rolls/Cs.
Dimensions (L x W) 3 1/2 x 1 in.
Length (English) 3.5 in.
Width (English) 1 in.
Width (Metric) 25.4 mm