Learn More
PDC Direct Thermal Paper Label Permanent, 1300/Roll, 8 Rolls/Case
Catalog No. 21235862
Change view
Click to view available options
Quantity:
1300/Roll, 8 Rolls/Cs.
Catalog No. Quantity
21-235-862 1300/Roll, 8 Rolls/Cs.
1 options
Catalog No. 21-235-862 Supplier PDC Supplier No. THERMD35
Add to Cart
Add to Cart
Compatible with Zebra, Intermec, and Datamax printers, these labels ensure durable adhesion, sharp printing, and versatile use.
  • Multi-purpose direct thermal labels engineered for reliable performance across demanding environments
  • Optimized for barcode clarity and accurate scanning in healthcare, logistics, and industrial settings
  • Strong permanent adhesive ensures long-lasting application even on challenging surfaces
  • Glove-friendly design suitable for sterile medical and laboratory environments
  • Perforated format with 1/8 in. gap allows quick, hassle-free label separation
  • High-volume packaging with 1300 labels per roll and 8 rolls per case supports bulk labeling needs
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1300/Roll, 8 Rolls/Cs.
Dimensions (L x W) 4 x 1 in.
Length (English) 1 in.
Width (English) 4 in.
Width (Metric) 101.6 mm