Learn More
PDC Direct Thermal Paper Label Permanent, 1500/Roll, 8 Rolls/Case
Catalog No. 21235847
Change view
Click to view available options
Quantity:
1500/Roll, 8 Rolls/Cs.
Catalog No. Quantity
21-235-847 1500/Roll, 8 Rolls/Cs.
1 options
Catalog No. 21-235-847 Supplier PDC Supplier No. THERMD13
Add to Cart
Add to Cart
Compatible with Zebra, Intermec, and Datamax printers, featuring strong adhesive, clear printing, and versatile multi-purpose use.
  • Permanent adhesive ensures durable label adhesion
  • Compatible with Zebra, Intermec, Datamax, and other printers
  • Perforated design allows easy peeling and smooth separation
  • Rounded corners help reduce curling and extend label lifespan
  • Glove-friendly adhesive available in permanent or removable options
  • Multi-purpose use suitable for healthcare, manufacturing, and logistics
  • High-quality direct thermal paper for crisp printing without ribbons
  • Designed to reduce printer jams and improve workflow efficiency
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1500/Roll, 8 Rolls/Cs.
Dimensions (L x W) 4 x 3 in.
Length (English) 4 in.
Width (English) 3 in.
Width (Metric) 76.2 mm