Learn More
PDC Direct Thermal Paper Label Permanent, 2500/Roll, 8 Rolls/Case
Catalog No. 21235846
Change view
Click to view available options
Quantity:
2500/Roll, 8 Rolls/Cs.
Catalog No. Quantity
21-235-846 2500/Roll, 8 Rolls/Cs.
1 options
Catalog No. 21-235-846 Supplier PDC Supplier No. THERMD11
Add to Cart
Add to Cart
Compatible with Zebra, Intermec, and Datamax printers, featuring strong adhesive, clear printing, and versatile use.
  • Permanent adhesive for secure, lasting label application
  • Compatible with Zebra, Intermec, Datamax, and similar printers
  • Perforated design allows easy peeling and smooth separation
  • Rounded corners help reduce peeling and curling
  • Glove-friendly adhesive available in permanent or removable options
  • Multi-purpose design suitable for healthcare, manufacturing, and logistics
  • High-quality direct thermal paper ensures clear barcode and text printing
  • Designed to minimize jams and improve printing efficiency
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 2500/Roll, 8 Rolls/Cs.
Dimensions (L x W) 4 x 2 in.
Length (English) 4 in.
Width (English) 2 in.
Width (Metric) 50.8 mm