Learn More
PDC Direct Thermal Paper Label Permanent, 500/Roll, 8 Rolls/Case
Catalog No. 21235860
Change view
Click to view available options
Quantity:
500/Roll, 8 Rolls/Cs.
Catalog No. Quantity
21-235-860 500/Roll, 8 Rolls/Cs.
1 options
Catalog No. 21-235-860 Supplier PDC Supplier No. THERMD29
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed for Eltron LP and TLP printers, with glove-friendly adhesive, perforations, and bulk packaging for easy labeling.
  • Compatible with Eltron LP, TLP Series, and other compatible printers
  • Durable, glove-friendly permanent adhesive for reliable labeling
  • Rounded corners prevent peeling and maintain neat edges
  • Perforated design allows fast and clean label separation
  • Direct thermal printing eliminates need for ribbons or ink
  • Rolls fit standard printers for smooth operation
  • Bulk pack provides continuous supply for frequent labeling
  • Suitable for inventory, shipping, and specimen labeling
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 500/Roll, 8 Rolls/Cs.
Dimensions (L x W) 4 x 3 in.
Length (English) 4 in.
Width (English) 3 in.
Width (Metric) 76.2 mm