Learn More
PDC Direct Thermal Paper Label Permanent, 735/Roll, 8 Rolls/Case
Catalog No. 21235855
Change view
Click to view available options
Quantity:
735/Roll, 8 Rolls/Cs.
Catalog No. Quantity
21-235-855 735/Roll, 8 Rolls/Cs.
1 options
Catalog No. 21-235-855 Supplier PDC Supplier No. THERMD22
Add to Cart
Add to Cart
Compatible with Eltron LP and TLP printers, these labels ensure strong adhesion, clear printing, and versatile use.
  • Compatible with Eltron LP, TLP Series, and other widely used printers for seamless integration
  • Permanent adhesive provides strong and durable labeling on different surfaces
  • Direct thermal printing eliminates the need for ink or ribbon, reducing maintenance
  • Designed for diverse applications including asset tracking, specimen labeling, and general use
  • Rounded corner design reduces peeling and curling, ensuring secure placement
  • Perforated format allows quick and easy label separation for efficient workflow
  • Optimized roll design supports standard printer compatibility for smooth operations
  • Glove-friendly adhesive makes application easier in healthcare, laboratory, and industrial environments
  • Bulk supply ensures consistent availability to meet high-volume labeling needs
  • Reliable performance trusted for accuracy and consistency across demanding tasks
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 735/Roll, 8 Rolls/Cs.
Dimensions (L x W) 3 x 2 in.
Length (English) 3 in.
Width (English) 2 in.
Width (Metric) 50.8 mm