Learn More
PDC Direct Thermal Paper Label Permanent, 735/Roll, 8 Rolls/Case
Catalog No. 21235859
Change view
Click to view available options
Quantity:
735/Roll, 8 Rolls/Cs.
Catalog No. Quantity
21-235-859 735/Roll, 8 Rolls/Cs.
1 options
Catalog No. 21-235-859 Supplier PDC Supplier No. THERMD27
Add to Cart
Add to Cart
Compatible with Eltron LP and TLP printers, featuring glove-friendly adhesive, perforations, and bulk packaging for versatile use.
  • Designed for use with Eltron LP, TLP Series, and other compatible printers for reliable performance
  • Permanent adhesive ensures durable and long-lasting label application on various surfaces
  • Rounded corners prevent peeling, maintaining a professional finish
  • Perforated layout allows smooth, easy label separation for efficient workflows
  • Glove-friendly adhesive supports safe and convenient use in healthcare and laboratory environments
  • Bulk packaging provides continuous supply to meet high-volume labeling needs
  • Versatile solution suitable for specimen labeling, inventory management, and asset tracking
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 735/Roll, 8 Rolls/Cs.
Dimensions (L x W) 4 x 2 in.
Length (English) 4 in.
Width (English) 2 in.
Width (Metric) 50.8 mm